2-Iodobenzonitrile ≥98.0%

Supplier: TCI America

I0659-1G I0659-25G
TCI0659-1GEA 36 USD
TCI0659-1G TCI0659-25G
2-Iodobenzonitrile ≥98.0%
2-Iodobenzonitrile

CAS Number: 4387-36-4
Molecular Formula: C7H4IN
Molecular Weight: 229.02
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 147
Melting point (°C): 55


Other Product Names / Synonyms: 1-Cyano-2-iodobenzene

Formula: C₇H₄IN
Boiling Pt: 147 °C
Melting Pt: 55 °C
MDL Number: MFCD00079761
CAS Number: 4387-36-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f4c09c8f2fb444a00d25bb4a7dde42404ff614d&vid=0f4c09c8f2fb444a00d25bb4a7dde42404ff614d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR