2,4,6-Trichlorophenyl isocyanate ≥98.0%

Supplier: TCI America

I0565-5G I0565-1G
TCI0565-005GEA 132 USD
TCI0565-005G TCI0565-1G
2,4,6-Trichlorophenyl isocyanate ≥98.0%
2,4,6-Trichlorophenyl isocyanate

CAS Number: 2505-31-9
MDL Number: MFCD00013845
Molecular Formula: C7H2Cl3NO
Molecular Weight: 222.45
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 101
Melting point (°C): 67
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Isocyanic Acid 2,4,6-Trichlorophenyl Ester

Formula: C₇H₂Cl₃NO
Boiling Pt: 101 °C
Melting Pt: 67 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00013845
CAS Number: 2505-31-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6fa394c68ac096996f600e001c9936a1401296e0&vid=6fa394c68ac096996f600e001c9936a1401296e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR