4-Bromophenyl isothiocyanate ≥98.0%

Supplier: TCI America

Synonyms: 1-Bromo-4-isothiocyanatobenzene, 4-Bromophenylisothiocyanate

I0526-5G I0526-25G
TCI0526-005GEA 184.1 USD
TCI0526-005G TCI0526-025G
4-Bromophenyl isothiocyanate ≥98.0%
4-Bromophenyl isothiocyanate

CAS Number: 1985-12-2
MDL Number: MFCD00004808
Molecular Formula: C7H4BrNS
Molecular Weight: 214.08
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 58


Other Product Names / Synonyms: Isothiocyanic Acid 4-Bromophenyl Ester

Formula: C₇H₄BrNS
MW: 214.09 g/mol
Boiling Pt: 142…144 °C (14 mmHg)
Melting Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00004808
CAS Number: 1985-12-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=985654f063832349cadcb644732b84588830df67&vid=985654f063832349cadcb644732b84588830df67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR