2-Methoxyphenyl isothiocyanate ≥98.0%

Supplier: TCI America

I0512-25G I0512-5G
TCI0512-025GEA 207.1 USD
TCI0512-025G TCI0512-005G
2-Methoxyphenyl isothiocyanate ≥98.0%
2-Methoxyphenyl isothiocyanate

CAS Number: 3288-04-8
MDL Number: MFCD00011675
Molecular Formula: C8H7NOS
Molecular Weight: 165.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 267
Flash Point (°C): 110
Specific Gravity (20/20): 1.19


Other Product Names / Synonyms: Isothiocyanic Acid 2-Methoxyphenyl Ester

Formula: C₈H₇NOS
Boiling Pt: 267 °C
Density: 1.19 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00011675
CAS Number: 3288-04-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f41b94bd5137b146c3a69ad402f3c68b102db80&vid=8f41b94bd5137b146c3a69ad402f3c68b102db80&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR