Hexyl isocyanate ≥98.0%

Supplier: TCI America

I0492-5ML I0492-25ML
TCI0492-005MLEA 137.53 USD
TCI0492-005ML TCI0492-025ML
Hexyl isocyanate ≥98.0%
Hexyl isocyanate

CAS Number: 2525-62-4
MDL Number: MFCD00037047
Molecular Formula: C7H13NO
Molecular Weight: 127.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 164
Flash Point (°C): 60
Specific Gravity (20/20): 0.88
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Isocyanic Acid Hexyl Ester

Formula: C₇H₁₃NO
Boiling Pt: 164 °C
Density: 0.88 (20 °C)
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00037047
CAS Number: 2525-62-4
EINECS: 219-763-8
UN: 3080
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec71c6ad8393cde11a6a24ce862f6d0a4b1e6cb3&vid=b3133c011b90b256667cc4592d5abec439bd5c30&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR