Butyl isobutyrate ≥98.0%

Supplier: TCI America

Synonyms: Isobutyric acid butyl ester

I0315-500ML I0315-25ML
TCI0315-500MLEA 152 USD
TCI0315-500ML TCI0315-025ML
Butyl isobutyrate ≥98.0%
Butyl isobutyrate

CAS Number: 97-87-0
MDL Number: MFCD00048773
Molecular Formula: C8H16O2
Molecular Weight: 144.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 150
Flash Point (°C): 43
Specific Gravity (20/20): 0.86


Other Product Names / Synonyms: Isobutyric Acid Butyl Ester

Formula: C₈H₁₆O₂
Boiling Pt: 150 °C
Density: 0.86 (20 °C)
Flash Pt: 43 °C
MDL Number: MFCD00048773
CAS Number: 97-87-0
EINECS: 202-614-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e592981e9020826ade6546c8180b8b3f63893611&vid=e592981e9020826ade6546c8180b8b3f63893611&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR