2-Methyl-4-undecanone ≥97.0%

Supplier: TCI America

Synonyms: Heptyl Isobutyl ketone

I0250-25G I0250-1G
TCI0250-025GEA 503 USD
TCI0250-025G TCI0250-1G
2-Methyl-4-undecanone ≥97.0%
2-Methyl-4-undecanone

CAS Number: 19594-40-2
MDL Number: MFCD00027319
Molecular Formula: C12H24O
Molecular Weight: 184.32
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 124
Flash Point (°C): 91
Specific Gravity (20/20): 0.82


Other Product Names / Synonyms: Heptyl Isobutyl Ketone

Formula: C₁₂H₂₄O
MW: 184.32 g/mol
Boiling Pt: 124 °C
Density: 0.82 (20 °C)
Flash Pt: 91 °C
MDL Number: MFCD00027319
CAS Number: 19594-40-2
EINECS: 243-176-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d977fc316ffd3bab4b404231e72c6f0a1027e972&vid=d977fc316ffd3bab4b404231e72c6f0a1027e972&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR