Itaconic anhydride ≥95.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Methylenesuccinic anhydride, 3-Methylenedihydrofuran-2,5-dione

I0203-25G I0203-500G I0203-100G
TCI0203-025GEA 31.53 USD
TCI0203-025G TCI0203-500G TCI0203-100G
Itaconic anhydride ≥95.0% (by titrimetric analysis)
Itaconic anhydride

CAS Number: 2170-03-8
MDL Number: MFCD00005530
Molecular Formula: C5H4O3
Molecular Weight: 112.08
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Boiling point (°C): 115
Melting point (°C): 68


Other Product Names / Synonyms: 3,4-Dihydro-3-methylene-2,5-furandione, Methylenesuccinic Anhydride

Formula: C₅H₄O₃
MW: 112.08 g/mol
Boiling Pt: 115 °C
Melting Pt: 68 °C
MDL Number: MFCD00005530
CAS Number: 2170-03-8
EINECS: 218-518-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ccc16490294f02cebd76b99559380f0644ffc1db&vid=ccc16490294f02cebd76b99559380f0644ffc1db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR