Octadecyl isocyanate ≥80.0%

Supplier: TCI America

Synonyms: Stearyl isocyanate, Isocyanic acid octadecyl ester, Isocyanic acid stearyl ester

I0130-250G I0130-25G
TCI0130-250GEA 140.38 USD
TCI0130-250G TCI0130-025G
Octadecyl isocyanate ≥80.0%
Octadecyl isocyanate

CAS Number: 112-96-9
MDL Number: MFCD00002041
Molecular Formula: C19H37NO
Molecular Weight: 295.51
Purity/Analysis Method: >80.0% (GC)
Form: Clear Liquid
Boiling point (°C): 220
Flash Point (°C): 202
Freezing point (°C): 19
Specific Gravity (20/20): 0.86
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Isocyanic Acid Octadecyl Ester, Isocyanic Acid Stearyl Ester, Stearyl Isocyanate

Formula: C₁₉H₃₇NO
MW: 295.51 g/mol
Boiling Pt: 220 °C
Melting Pt: 15…16 °C
Density: 0.86 (20 °C)
Flash Pt: 202 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002041
CAS Number: 112-96-9
EINECS: 204-019-7
UN: 2206
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f731cadd6bcde4827f9f04d7130d1e1389df09e0&vid=f731cadd6bcde4827f9f04d7130d1e1389df09e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >80.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR