3-Iodo-L-tyrosine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: H-Tyr(3-I)-OH, (S)-2-Amino-3-(4-hydroxy-3-iodophenyl)propanoic acid

I0075-1G I0075-5G
TCI0075-001GEA 78 USD
TCI0075-001G TCI0075-5G
3-Iodo-L-tyrosine ≥98.0% (by titrimetric analysis)
3-Iodo-L-tyrosine

CAS Number: 70-78-0
MDL Number: MFCD00002608
Molecular Formula: C9H10INO3
Molecular Weight: 307.09
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 199
Specific rotation [a]20/D: -3 deg (C=2, 1mol/L Hcl)
Storage Temperature: 0-10°C


Product Categories: Bioscience, Biologically-modified Amino Acids
Other Product Names / Synonyms: 3-Monoiodo-L-tyrosine, H-Tyr(3-I)-OH

Formula: C₉H₁₀INO₃
MW: 307.1 g/mol
Melting Pt: 199 °C
MDL Number: MFCD00002608
CAS Number: 70-78-0
EINECS: 200-744-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a26782b60b8c57594d7f41bb26d232e0e7d29057&vid=a26782b60b8c57594d7f41bb26d232e0e7d29057&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR