3-Hexylthiophene ≥98.0%

Supplier: TCI America

H0756-5G H0756-1G H0756-25G
TCH0756-005GEA 100 USD
TCH0756-005G TCH0756-001G TCH0756-25G
3-Hexylthiophene ≥98.0%
3-Hexylthiophene

CAS Number: 1693-86-3
MDL Number: MFCD00143181
Molecular Formula: C10H16S
Molecular Weight: 168.30
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 103
Flash Point (°C): 37
Specific Gravity (20/20): 0.94

Formula: C₁₀H₁₆S
MW: 168.3 g/mol
Boiling Pt: 103 °C
Density: 0.94 (20 °C)
Flash Pt: 37 °C
Storage Temperature: Ambient
MDL Number: MFCD00143181
CAS Number: 1693-86-3
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d94bdca233ba43d7cb3cea1a79790c479b8ad4bb&vid=d94bdca233ba43d7cb3cea1a79790c479b8ad4bb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR