5-Hydroxy-L-tryptophan ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
(S)-2-Amino-3-(5-hydroxy-1H-indol-3-yl)propanoic acid, 5-Hydroxyl-L-tryptophan, (-)-5-Hydroxy-L-Tryptophane, L-5-Hydroxytryptophan, Oxitriptan, H-Trp(5-OH)-OH
H0531-5G
H0531-1G
TCH0531-005GEA
63.16
USD
TCH0531-005G
TCH0531-001G
5-Hydroxy-L-tryptophan ≥98.0% (by HPLC, titration analysis)
5-Hydroxy-L-tryptophan
CAS Number: 4350-09-8
MDL Number: MFCD00064341
Molecular Formula: C11H12N2O3
Molecular Weight: 220.23
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: -33 deg (C=1, H2O)
Product Categories: Bioscience, Biologically-modified Amino Acids
Other Product Names / Synonyms: H-Trp(5-OH)-OH
Formula:
C₁₁H₁₂N₂O₃ MW: 220.23 g/mol Melting Pt: 273 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00064341 CAS Number: 4350-09-8 EINECS: 224-411-1 UN: 2811 ADR: 6.1,III Merck Index: 12,04895 |
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...