cis-4-Heptenal ≥95.0%
Supplier: TCI America
CAS Number: 6728-31-0
MDL Number: MFCD00014682
Molecular Formula: C7H12O
Molecular Weight: 112.17
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 58
Flash Point (°C): 44
Specific Gravity (20/20): 0.85
Storage Temperature: 0-10°C
Formula:
C₇H₁₂O Boiling Pt: 58 °C Density: 0.85 (20 °C) Flash Pt: 44 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014682 CAS Number: 6728-31-0 EINECS: 229-779-7 UN: 1989 ADR: 3,III |
|
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...