cis-2-Heptene ≥95.0%
Supplier: TCI America
Synonyms:
(Z)-2-Heptene
H0411-1ML
H0411-10ML
TCH0411-001MLEA
68
USD
TCH0411-001ML
TCH0411-010ML
cis-2-Heptene ≥95.0%
cis-2-Heptene
CAS Number: 6443-92-1
MDL Number: MFCD00063976
Molecular Formula: C7H14
Molecular Weight: 98.19
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 99
Flash Point (°C): -6
Specific Gravity (20/20): 0.71
Formula:
C₇H₁₄ MW: 98.19 g/mol Boiling Pt: 99 °C Melting Pt: –110 °C Density: 0.71 (20 °C) Flash Pt: –6 °C |
MDL Number:
MFCD00063976 CAS Number: 6443-92-1 EINECS: 229-242-7 UN: 3295 ADR: 3,II |
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...