cis-3-Hexene ≥97.0%

Supplier: TCI America

H0398-1ML H0398-5ML
TCH0398-001MLEA 30.6 USD
TCH0398-001ML TCH0398-005ML
cis-3-Hexene ≥97.0%
cis-3-Hexene

CAS Number: 7642-09-3
MDL Number: MFCD00085300
Molecular Formula: C6H12
Molecular Weight: 84.16
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 67
Flash Point (°C): -25
Specific Gravity (20/20): 0.68

Formula: C₆H₁₂
Boiling Pt: 67 °C
Density: 0.68 (20 °C)
Flash Pt: -25 °C
MDL Number: MFCD00085300
CAS Number: 7642-09-3

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR