N-(2-Hydroxyethyl)phthalimide ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
2-(2-Hydroxyethyl)isoindoline-1,3-dione
H0245-25G
H0245-500G
TCH0245-025GEA
40.8
USD
TCH0245-025G
TCH0245-500G
N-(2-Hydroxyethyl)phthalimide ≥98.0% (by HPLC, total nitrogen)
N-(2-Hydroxyethyl)phthalimide
CAS Number: 3891-07-4
MDL Number: MFCD00005903
Molecular Formula: C10H9NO3
Molecular Weight: 191.19
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 128
Formula:
C₁₀H₉NO₃ MW: 191.19 g/mol Melting Pt: 128 °C |
MDL Number:
MFCD00005903 CAS Number: 3891-07-4 EINECS: 223-434-4 |
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,N) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...