4-Fluoro-2-iodoaniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

F0706-25G F0706-5G
TCF0706-25GEA 169.5 USD
TCF0706-25G TCF0706-5G
4-Fluoro-2-iodoaniline ≥98.0% (by GC, titration analysis)
4-Fluoro-2-iodoaniline

CAS Number: 61272-76-2
MDL Number: MFCD00042584
Molecular Formula: C6H5FIN
Molecular Weight: 237.02
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Specific Gravity (20/20): 1.99

Formula: C₆H₅FIN
MW: 237.02 g/mol
Density: 1.99 (20 °C)
MDL Number: MFCD00042584
CAS Number: 61272-76-2
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=21f64be812ce69302624ce7dcd45d4d5087ef50b&vid=21f64be812ce69302624ce7dcd45d4d5087ef50b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR