5-Fluoro-2-nitroanisole ≥98.0%
Supplier: TCI America
Synonyms:
4-Fluoro-2-methoxy-1-nitrobenzene
F0703-25G
F0703-5G
TCF0703-25GEA
62
USD
TCF0703-25G
TCF0703-5G
5-Fluoro-2-nitroanisole ≥98.0%
5-Fluoro-2-nitroanisole
CAS Number: 448-19-1
MDL Number: MFCD00077541
Molecular Formula: C7H6FNO3
Molecular Weight: 171.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 272
Melting point (°C): 49
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Fluoro-2-methoxy-1-nitrobenzene
Formula:
C₇H₆FNO₃ MW: 171.13 g/mol Boiling Pt: 272 °C Melting Pt: 49 °C Storage Temperature: Ambient |
MDL Number:
MFCD00077541 CAS Number: 448-19-1 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...