6-Fluoronicotinic acid ≥98.0%

Supplier: TCI America

Synonyms: 2-Fluoro-5-pyridinecarboxylic acid

F0695-1G F0695-200MG F0695-5G
TCF0695-1GEA 116 USD
TCF0695-1G TCF0695-200MG TCF0695-5G
6-Fluoronicotinic acid ≥98.0%
6-Fluoronicotinic acid

CAS Number: 403-45-2
MDL Number: MFCD01859863
Molecular Formula: C6H4FNO2
Molecular Weight: 141.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 147


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 6-Fluoropyridine-3-carboxylic Acid

Formula: C₆H₄FNO₂
MW: 141.1 g/mol
Melting Pt: 147 °C
MDL Number: MFCD01859863
CAS Number: 403-45-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=731777c53ad4979e52168a5e559baf4685a82586&vid=731777c53ad4979e52168a5e559baf4685a82586&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR