2-Fluoro-4-nitroanisole ≥98.0%

Supplier: TCI America
Warning

F0617-5G F0617-1G
TCF0617-5GEA 65 USD
TCF0617-5G TCF0617-1G
2-Fluoro-4-nitroanisole ≥98.0%
2-Fluoro-4-nitroanisole

CAS Number: 455-93-6
MDL Number: MFCD00061095
Molecular Formula: C7H6FNO3
Molecular Weight: 171.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 105


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Fluoro-1-methoxy-4-nitrobenzene

Formula: C₇H₆FNO₃
MW: 171.13 g/mol
Melting Pt: 105 °C
Storage Temperature: Ambient
MDL Number: MFCD00061095
CAS Number: 455-93-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=500a2e3628eba12fa7368dba8d1ee4b04735e29d&vid=500a2e3628eba12fa7368dba8d1ee4b04735e29d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR