3-Fluoro-p-cresol ≥98.0%
Supplier: TCI America
Synonyms:
3-Fluoro-4-methylphenol, 1-Fluoro-3-hydroxy-6-methylbenzene
F0481-1G
F0481-5G
TCF0481-1GEA
33
USD
TCF0481-1G
TCF0481-5G
3-Fluoro-p-cresol ≥98.0%
3-Fluoro-p-cresol
CAS Number: 452-78-8
MDL Number: MFCD00143164
Molecular Formula: C7H7FO
Molecular Weight: 126.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 196
Freezing point (°C): 29
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-Fluoro-4-methylphenol
Formula:
C₇H₇FO Boiling Pt: 196 °C |
MDL Number:
MFCD00143164 CAS Number: 452-78-8 UN: 3265 ADR: 8,III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...