4-Fluoro-3-methoxybenzonitrile

Supplier: TCI America

F0408-1G F0408-5G
TCF0408-001GEA 25 USD
TCF0408-001G TCF0408-005G
4-Fluoro-3-methoxybenzonitrile
4-Fluoro-3-methoxybenzonitrile

CAS Number: 243128-37-2
MDL Number: MFCD01569371
Molecular Formula: C8H6FNO
Molecular Weight: 151.14
Form: Crystal
Color: White
Melting point (°C): 109

Formula: C₈H₆FNO
MW: 151.14 g/mol
Melting Pt: 109 °C
MDL Number: MFCD01569371
CAS Number: 243128-37-2
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=30b3e05ff16d86e86e51a11013fefd6c370024ca&vid=30b3e05ff16d86e86e51a11013fefd6c370024ca&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR