3-Fluorocinnamic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: 3-(3-Fluorophenyl)acrylic acid, m-Fluorocinnamic acid, 3-Fluorocinnamic Acid

F0264-5G F0264-25G
TCF0264-005GEA 66 USD
TCF0264-005G TCF0264-25G
3-Fluorocinnamic acid ≥98.0% (by titrimetric analysis)
3-Fluorocinnamic acid

CAS Number: 458-46-8
MDL Number: MFCD00004383
Molecular Formula: C9H7FO2
Molecular Weight: 166.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 166

Formula: C9H7FO2
MW: 166.15 g/mol
Melting Pt: 166 °C
MDL Number: MFCD00004383
CAS Number: 458-46-8
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c854c9e935375bc4622423b17367bda8c7515f6&vid=1c854c9e935375bc4622423b17367bda8c7515f6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR