Furfuryl mercaptan ≥98.0%

Supplier: TCI America

Synonyms: 2-Furanmethanethiol, 2-Furanmethanthiol, 2-Furylmethanethiol

F0077-25G F0077-500G
TCF0077-025GEA 47.79 USD
TCF0077-025G TCF0077-500G
Furfuryl mercaptan ≥98.0%
Furfuryl mercaptan

CAS Number: 98-02-2
MDL Number: MFCD00003254
Molecular Formula: C5H6OS
Molecular Weight: 114.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 155
Flash Point (°C): 45
Specific Gravity (20/20): 1.13

Formula: C₅H₆OS
MW: 114.17 g/mol
Boiling Pt: 155 °C
Density: 1.13 (20 °C)
Flash Pt: 45 °C
MDL Number: MFCD00003254
CAS Number: 98-02-2
EINECS: 202-628-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f79d1db0282025769a1b5fefc8a8e3234751b31d&vid=f79d1db0282025769a1b5fefc8a8e3234751b31d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR