Ethyl-10-undecenoate ≥97.0%

Supplier: TCI America

Synonyms: 10-Undecenoic acid ethyl ester

E0771-250G E0771-25G
TCE0771-250GEA 187 USD
TCE0771-250G TCE0771-25G
Ethyl-10-undecenoate ≥97.0%
Ethyl-10-undecenoate

CAS Number: 692-86-4
MDL Number: MFCD00009220
Molecular Formula: C13H24O2
Molecular Weight: 212.33
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 259
Melting point (°C): -38
Flash Point (°C): 110
Specific Gravity (20/20): 0.88


Other Product Names / Synonyms: 10-Undecenoic Acid Ethyl Ester

Formula: C₁₃H₂₄O₂
Boiling Pt: 259 °C
Melting Pt: -38 °C
Density: 0.88 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00009220
CAS Number: 692-86-4
EINECS: 211-734-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b28b4f239ceed279624fdefbcca1cab9fdb8b836&vid=b28b4f239ceed279624fdefbcca1cab9fdb8b836&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR