Erythromycin ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
(-)-Erythromycin
E0751-25G
E0751-5G
TCE0751-25GEA
155.28
USD
TCE0751-25G
TCE0751-5G
Erythromycin ≥98.0% (by titrimetric analysis)
Erythromycin
CAS Number: 114-07-8
MDL Number: MFCD00084654
Molecular Formula: C37H67NO13
Molecular Weight: 733.94
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: -74 deg (C=2, EtOH)
Formula:
C₃₇H₆₇NO₁₃ MW: 733.95 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00084654 CAS Number: 114-07-8 EINECS: 204-040-1 |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...