Ethosuximide ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2-Ethyl-2-methylsuccinimide, 3-Ethyl-3-methylpyrrolidine-2,5-dione
E0746-25G
E0746-5G
TCE0746-25GEA
124
USD
TCE0746-25G
TCE0746-5G
Ethosuximide ≥98.0% (by GC, titration analysis)
Ethosuximide
CAS Number: 77-67-8
MDL Number: MFCD00072123
Molecular Formula: C7H11NO2
Molecular Weight: 141.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 150
Melting point (°C): 51
Storage Temperature: 0-10°C
Product Categories: Bioscience, Neurotransmitter Agent Ingredients, Voltage-dependent Calcium Channel (VDCC) Antagonists
Other Product Names / Synonyms: 2-Ethyl-2-methylsuccinimide
Formula:
C₇H₁₁NO₂ MW: 141.17 g/mol Boiling Pt: 150 °C Melting Pt: 51 °C |
MDL Number:
MFCD00072123 CAS Number: 77-67-8 EINECS: 201-048-7 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...