2-Ethyl-2-methyl-1,3-dioxolane ≥98.0%
Supplier: TCI America
E0515-25ML
E0515-500ML
TCE0515-025MLEA
369
USD
TCE0515-025ML
TCE0515-500ML
2-Ethyl-2-methyl-1,3-dioxolane ≥98.0%
2-Ethyl-2-methyl-1,3-dioxolane
CAS Number: 126-39-6
MDL Number: MFCD00014108
Molecular Formula: C6H12O2
Molecular Weight: 116.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 117
Flash Point (°C): 13
Specific Gravity (20/20): 0.94
Formula:
C₆H₁₂O₂ MW: 116.16 g/mol Boiling Pt: 117 °C Density: 0.94 (20 °C) Flash Pt: 13 °C |
MDL Number:
MFCD00014108 CAS Number: 126-39-6 EINECS: 204-785-2 UN: 1993 ADR: 3,I |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...