3-Ethylthiophene ≥98.0%

Supplier: TCI America

E0378-5ML E0378-25ML
TCE0378-005MLEA 193 USD
TCE0378-005ML TCE0378-025ML
3-Ethylthiophene ≥98.0%
3-Ethylthiophene

CAS Number: 1795-01-3
MDL Number: MFCD00070514
Molecular Formula: C6H8S
Molecular Weight: 112.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 141
Specific Gravity (20/20): 1.00

Formula: C₆H₈S
Boiling Pt: 141 °C
Density: 1.00 (20 °C)
MDL Number: MFCD00070514
CAS Number: 1795-01-3
EINECS: 217-267-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be8a63787f6053f0833b830246d0aff4edb73309&vid=be8a63787f6053f0833b830246d0aff4edb73309&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR