4-Ethyl-m-xylene ≥95.0%

Supplier: TCI America

E0241-1ML E0241-5ML
TCE0241-001MLEA 137 USD
TCE0241-001ML TCE0241-5ML
4-Ethyl-m-xylene ≥95.0%
4-Ethyl-m-xylene

CAS Number: 874-41-9
MDL Number: MFCD00039905
Molecular Formula: C10H14
Molecular Weight: 134.22
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 186
Specific Gravity (20/20): 0.88


Other Product Names / Synonyms: 2,4-Dimethylethylbenzene, 1-Ethyl-2,4-dimethylbenzene

Formula: C₁₀H₁₄
Boiling Pt: 186 °C
Melting Pt: -62.9 °C
Density: 0.88 (20 °C)
MDL Number: MFCD00039905
CAS Number: 874-41-9
EINECS: 212-860-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac8cfb157d5a4bf7bb6c3ff10679aa31eaa9021c&vid=ac8cfb157d5a4bf7bb6c3ff10679aa31eaa9021c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR