N-Ethyl-m-toluidine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
N-Ethyl-3-methylaniline, 3-(Ethylamino)toluene
E0178-25ML
E0178-500ML
TCE0178-025MLEA
37
USD
TCE0178-025ML
TCE0178-500ML
N-Ethyl-m-toluidine ≥98.0% (by GC, titration analysis)
N-Ethyl-m-toluidine
CAS Number: 102-27-2
MDL Number: MFCD00009027
Molecular Formula: C9H13N
Molecular Weight: 135.21
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 221
Flash Point (°C): 104
Specific Gravity (20/20): 0.95
Other Product Names / Synonyms: N-Ethyl-3-methylaniline, 3-(Ethylamino)toluene
Formula:
C₉H₁₃N MW: 135.21 g/mol Boiling Pt: 221 °C Melting Pt: 9 °C Density: 0.95 (20 °C) Flash Pt: 104 °C |
MDL Number:
MFCD00009027 CAS Number: 102-27-2 EINECS: 203-019-4 UN: 2754 ADR: 6.1,II |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...