Ethylenediamine monohydrate ≥98.0%

Supplier: TCI America
Danger

Synonyms: 1,2-Diaminoethane monohydrate, 1,2-Ethanediamine monohydrate

E0081-25ML E0081-500ML
TCE0081-025MLEA 40.2 USD
TCE0081-025ML TCE0081-500ML
Ethylenediamine monohydrate ≥98.0%
Ethylenediamine monohydrate

CAS Number: 6780-13-8
MDL Number: MFCD00149563
Molecular Formula: C2H8N2
Molecular Weight: 60.10
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 118
Melting point (°C): 10
Flash Point (°C): 34
Specific Gravity (20/20): 0.96

Formula: C₂H₈N₂·1H₂O
MW: 78.11 g/mol
Boiling Pt: 118
Melting Pt: 11
Density: 0.96 (20 °C)
Flash Pt: 34 °C
Storage Temperature: Ambient
MDL Number: MFCD00149563
CAS Number: 6780-13-8
EINECS: 203-468-6
UN: 1604
ADR: 8 II
Merck Index: 13,03829
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89aa80e70251c02a34b7fca48373af16517f8eef&vid=89aa80e70251c02a34b7fca48373af16517f8eef&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR