2,3-Difluoropyridine ≥98.0%

Supplier: TCI America
Danger

D3892-5G D3892-1G
TCD3892-5GEA 328.5 USD
TCD3892-5G TCD3892-1G
2,3-Difluoropyridine ≥98.0%
2,3-Difluoropyridine

CAS Number: 1513-66-2
MDL Number: MFCD04114143
Molecular Formula: C5H3F2N
Molecular Weight: 115.08
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 126
Flash Point (°C): 31
Specific Gravity (20/20): 1.27

Formula: C₅H₃F₂N
MW: 115.08 g/mol
Boiling Pt: 126 °C
Density: 1.27 (20 °C)
Flash Pt: 31 °C
MDL Number: MFCD04114143
CAS Number: 1513-66-2
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=93b916641e56415bfbfb7bcb50fb15dad20b70f9&vid=93b916641e56415bfbfb7bcb50fb15dad20b70f9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR