4,4''-Diiodo-p-terphenyl ≥98.0%

Supplier: TCI America

D3534-1G D3534-5G
TCD3534-1GEA 137 USD
TCD3534-1G TCD3534-5G
4,4''-Diiodo-p-terphenyl ≥98.0%
4,4''-Diiodo-p-terphenyl

CAS Number: 19053-14-6
Molecular Formula: C18H12I2
Molecular Weight: 482.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 328

Formula: C₁₈H₁₂I₂
Melting Pt: 328 °C
CAS Number: 19053-14-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c08af0f426d73b5f8e01fb7ba569cb9e1fd0ef07&vid=c08af0f426d73b5f8e01fb7ba569cb9e1fd0ef07&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR