3,5-Dichlorothioanisole ≥98.0%
Supplier: TCI America
D3520-25G
D3520-5G
TCD3520-25GEA
88
USD
TCD3520-25G
TCD3520-5G
3,5-Dichlorothioanisole ≥98.0%
3,5-Dichlorothioanisole
CAS Number: 68121-46-0
MDL Number: MFCD00870149
Molecular Formula: C7H6Cl2S
Molecular Weight: 193.09
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 145
Flash Point (°C): 110
Specific Gravity (20/20): 1.36
Other Product Names / Synonyms: 1,3-Dichloro-5-(methylthio)benzene
Formula:
C₇H₆Cl₂S MW: 193.1 g/mol Boiling Pt: 145 °C Density: 1.36 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00870149 CAS Number: 68121-46-0 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...