4,4''-Dibromo-p-terphenyl ≥95.0%

Supplier: TCI America

Synonyms: 4,4''-Dibromo-1,1':4',1''-terphenyl

D3372-1G D3372-5G
TCD3372-1GEA 35 USD
TCD3372-1G TCD3372-5G
4,4''-Dibromo-p-terphenyl ≥95.0%
4,4''-Dibromo-p-terphenyl

CAS Number: 17788-94-2
Molecular Formula: C18H12Br2
Molecular Weight: 388.10
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 315

Formula: C₁₈H₁₂Br₂
MW: 388.1000 g/mol
Melting Pt: 315 °C
MDL Number: MFCD00272889
CAS Number: 17788-94-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d5843a99199de7713df47fd8146d7e7e29f4b51c&vid=d5843a99199de7713df47fd8146d7e7e29f4b51c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR