2,6-Dichloroisonicotinic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
CAS Number: 5398-44-7
Molecular Formula: C6H3Cl2NO2
Molecular Weight: 192.00
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 211
Other Product Names / Synonyms: 2,6-Dichloropyridine-4-carboxylic Acid
Formula:
C₆H₃Cl₂NO₂ Melting Pt: 211 °C |
MDL Number:
MFCD00234147 CAS Number: 5398-44-7 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...