2,5-Dichloroisonicotinic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2,5-Dichloropyridine-4-carboxylic acid

D2599-5G D2599-500MG
TCD2599-5GEA 387 USD
TCD2599-5G TCD2599-500MG
2,5-Dichloroisonicotinic acid ≥98.0% (by GC, titration analysis)
2,5-Dichloroisonicotinic acid

CAS Number: 88912-26-9
MDL Number: MFCD00466640
Molecular Formula: C6H3Cl2NO2
Molecular Weight: 192.00
Purity/Analysis Method: >98.0% (T,GC)
Form: Crystal


Other Product Names / Synonyms: 2,5-Dichloropyridine-4-carboxylic Acid

Formula: C₆H₃Cl₂NO₂
MW: 192 g/mol
MDL Number: MFCD00466640
CAS Number: 88912-26-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2fc6aa75d052ac516dd5c304ce3f5bac27b7d4e&vid=f2fc6aa75d052ac516dd5c304ce3f5bac27b7d4e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T,GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR