3,5-Dinitroaniline ≥98.0%

Supplier: TCI America

D2458-5G D2458-25G
TCD2458-005GEA 133.9 USD
TCD2458-005G TCD2458-025G
3,5-Dinitroaniline ≥98.0%
3,5-Dinitroaniline

CAS Number: 618-87-1
MDL Number: MFCD00007263
Molecular Formula: C6H5N3O4
Molecular Weight: 183.12
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 162

Formula: C₆H₅N₃O₄
MW: 183.12 g/mol
Melting Pt: 162 °C
Density: 1.601 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007263
CAS Number: 618-87-1
EINECS: 210-567-8
UN: 1596
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38df2df028708e1243ffe535ec6680f956104949&vid=7c3d8eb9929f90f93923942b2c35694518692b30&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR