2,2-Dimethylvaleric acid ≥98.0%
Supplier: TCI America
Synonyms:
2,2-Dimethylpentanoic acid
D2320-5ML
D2320-25ML
TCD2320-005MLEA
205
USD
TCD2320-005ML
TCD2320-25ML
2,2-Dimethylvaleric acid ≥98.0%
2,2-Dimethylvaleric acid
CAS Number: 1185-39-3
MDL Number: MFCD00021686
Molecular Formula: C7H14O2
Molecular Weight: 130.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 110
Flash Point (°C): 110
Specific Gravity (20/20): 0.92
Other Product Names / Synonyms: 2,2-Dimethylpentanoic Acid
Formula:
C₇H₁₄O₂ Boiling Pt: 110 °C Density: 0.92 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00021686 CAS Number: 1185-39-3 EINECS: 214-682-4 UN: 3265 ADR: 8,II |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...