1,3-Diaminopentane ≥96.0%

Supplier: TCI America

Synonyms: 1,3-Pentanediamine

D2252-25ML D2252-500ML
TCD2252-025MLEA 23 USD
TCD2252-025ML TCD2252-500ML
1,3-Diaminopentane ≥96.0%
1,3-Diaminopentane

CAS Number: 589-37-7
MDL Number: MFCD00134550
Molecular Formula: C5H14N2
Molecular Weight: 102.18
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 164
Flash Point (°C): 59
Specific Gravity (20/20): 0.86


Other Product Names / Synonyms: 1,3-Pentanediamine

Formula: C₅H₁₄N₂
MW: 102.18 g/mol
Boiling Pt: 164 °C
Density: 0.86 (20 °C)
Flash Pt: 59 °C
MDL Number: MFCD00134550
CAS Number: 589-37-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a2d7565d5f97e26fff01dc03b195476e75edfa9&vid=2a2d7565d5f97e26fff01dc03b195476e75edfa9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR