2,3-Dimethyl-2,3-dinitrobutane ≥98.0%
Supplier: TCI America
|
Danger
|
D1822-10G
D1822-25G
TCD1822-010GEA
87.07
USD
TCD1822-010G
TCD1822-25G
2,3-Dimethyl-2,3-dinitrobutane ≥98.0%
2,3-Dimethyl-2,3-dinitrobutane
CAS Number: 3964-18-9
MDL Number: MFCD00007392
Molecular Formula: C6H12N2O4
Molecular Weight: 176.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 215
Formula:
C₆H₁₂N₂O₄ MW: 176.17 g/mol Melting Pt: 215 °C Density: 1.34 g/cm³ (20 °C) |
MDL Number:
MFCD00007392 CAS Number: 3964-18-9 EINECS: 223-569-9 UN: 2811 ADR: 6.1,II |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...