Dimethyldiphenylsilane ≥97.0%

Supplier: TCI America

D1810-5ML D1810-25ML
TCD1810-005MLEA 106 USD
TCD1810-005ML TCD1810-025ML
Dimethyldiphenylsilane ≥97.0%
Dimethyldiphenylsilane

CAS Number: 778-24-5
MDL Number: MFCD00041326
Molecular Formula: C14H16Si
Molecular Weight: 212.37
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 118
Flash Point (°C): 100
Specific Gravity (20/20): 0.99

Formula: C₁₄H₁₆Si
Boiling Pt: 118 °C
Density: 0.99 (20 °C)
Flash Pt: 100 °C
MDL Number: MFCD00041326
CAS Number: 778-24-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62cfbf12f04f67580c5e9d83f71d5dbe77b063f4&vid=62cfbf12f04f67580c5e9d83f71d5dbe77b063f4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR