cis-1,4-Dimethylcyclohexane ≥98.0%

Supplier: TCI America

Synonyms: Cis-1,4-dimethylcyclohexane

D1715-5ML D1715-25ML
TCD1715-005MLEA 76 USD
TCD1715-005ML TCD1715-025ML
cis-1,4-Dimethylcyclohexane ≥98.0%
cis-1,4-Dimethylcyclohexane

CAS Number: 624-29-3
MDL Number: MFCD00064174
Molecular Formula: C8H16
Molecular Weight: 112.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 125
Flash Point (°C): 6
Specific Gravity (20/20): 0.78

Formula: C₈H₁₆
MW: 112.22 g/mol
Boiling Pt: 125 °C
Melting Pt: –42 °C
Density: 0.78 (20 °C)
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00064174
CAS Number: 624-29-3
EINECS: 210-840-1
UN: 2263
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d3633b1a4e98c68df7e93cf84a2a946a23cc4fe&vid=6d3633b1a4e98c68df7e93cf84a2a946a23cc4fe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR