Dichloromethyl methyl ether ≥97.0%

Supplier: TCI America

Synonyms: 1,1-Dichlorodimethyl ether

D1645-25G D1645-250G
TCD1645-025GEA 43.75 USD
TCD1645-025G TCD1645-250G
Dichloromethyl methyl ether ≥97.0%
Dichloromethyl methyl ether

CAS Number: 4885-02-3
MDL Number: MFCD00000828
Molecular Formula: C2H4Cl2O
Molecular Weight: 114.95
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 85
Flash Point (°C): 22
Specific Gravity (20/20): 1.30


Product Categories: Synthetic Organic Chemistry, Formylation, Halogenation
Other Product Names / Synonyms: asym-Dichlorodimethyl Ether

Formula: C₂H₄Cl₂O
MW: 114.96 g/mol
Boiling Pt: 85 °C
Density: 1.30 (20 °C)
Flash Pt: 22 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000828
CAS Number: 4885-02-3
EINECS: 225-498-9
UN: 2924
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=93fc349ff296d0c66266e11c6ecd87c40e1d8004&vid=68027ceeb92801d5a47f671890630f7e68842c34&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR