3,3-Dimethyl-1-butanol ≥95.0%

Supplier: TCI America

Synonyms: 3,3-Dimethylbutan-1-ol

D1333-5ML D1333-25ML
TCD1333-005MLEA 37 USD
TCD1333-005ML TCD1333-25ML
3,3-Dimethyl-1-butanol ≥95.0%
3,3-Dimethyl-1-butanol

CAS Number: 624-95-3
MDL Number: MFCD00002928
Molecular Formula: C6H14O
Molecular Weight: 102.18
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 142
Flash Point (°C): 56
Specific Gravity (20/20): 0.81


Other Product Names / Synonyms: Neohexanol

Formula: C₆H₁₄O
MW: 102.18 g/mol
Boiling Pt: 142 °C
Melting Pt: –65…–60 °C
Density: 0.81 (20 °C)
Flash Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD00002928
CAS Number: 624-95-3
EINECS: 210-872-6
UN: 1987
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4bb5f193056a46735bf249872c034be577d59553&vid=4bb5f193056a46735bf249872c034be577d59553&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR