1,3-Dioxane ≥98.0%

Supplier: TCI America

Synonyms: m-Dioxane, 1,3-Dioxacyclohexane

D0859-500ML D0859-25ML D0859-100ML
TCD0859-500MLEA 903.9 USD
TCD0859-500ML TCD0859-025ML TCD0859-100ML
1,3-Dioxane ≥98.0%
1,3-Dioxane

CAS Number: 505-22-6
MDL Number: MFCD00006566
Molecular Formula: C4H8O2
Molecular Weight: 88.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 105
Flash Point (°C): 1
Specific Gravity (20/20): 1.04

Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 105 °C
Melting Pt: –74.2 °C
Density: 1.04 (20 °C)
Flash Pt: 1 °C
Storage Temperature: Ambient
MDL Number: MFCD00006566
CAS Number: 505-22-6
EINECS: 208-005-1
UN: 1165
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f9a2189056255e552d54252f16c1eb012d0d440&vid=9f9a2189056255e552d54252f16c1eb012d0d440&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR