2,4-Dinitroaniline ≥98.0%
Supplier: TCI America
|
Danger
|
D0814-500G
D0814-25G
TCD0814-500GEA
93
USD
TCD0814-500G
TCD0814-025G
2,4-Dinitroaniline ≥98.0%
2,4-Dinitroaniline
CAS Number: 97-02-9
MDL Number: MFCD00007151
Molecular Formula: C6H5N3O4
Molecular Weight: 183.12
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 180
Flash Point (°C): 229
Formula:
C₆H₅N₃O₄ Boiling Pt: 401 °C (1013 hPa) Melting Pt: 180 °C Density: 1.6 g/cm³ (20 °C) Flash Pt: 229 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007151 CAS Number: 97-02-9 EINECS: 202-553-5 UN: 1596 ADR: 6.1,II Merck Index: 13,03298 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...