N,N-Dimethylpropionamide ≥98.0%

Supplier: TCI America

D0793-500ML D0793-25ML D0793-100ML
TCD0793-500MLEA 419 USD
TCD0793-500ML TCD0793-025ML TCD0793-100ML
N,N-Dimethylpropionamide ≥98.0%
N,N-Dimethylpropionamide

CAS Number: 758-96-3
MDL Number: MFCD00009301
Molecular Formula: C5H11NO
Molecular Weight: 101.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 176
Flash Point (°C): 62
Specific Gravity (20/20): 0.93

Formula: C₅H₁₁NO
MW: 101.15 g/mol
Boiling Pt: 176 °C
Density: 0.93 (20 °C)
Flash Pt: 62 °C
MDL Number: MFCD00009301
CAS Number: 758-96-3
EINECS: 212-064-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85054c74239e99b493b7267d3cc5a976e1d3d970&vid=85054c74239e99b493b7267d3cc5a976e1d3d970&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR