N,N-Dimethylglycine ethyl ester ≥98.0%
Supplier: TCI America
Synonyms:
Di-Me-Gly-OEt
D0726-25G
D0726-500G
TCD0726-025GEA
123
USD
TCD0726-025G
TCD0726-500G
N,N-Dimethylglycine ethyl ester ≥98.0%
N,N-Dimethylglycine ethyl ester
CAS Number: 33229-89-9
MDL Number: MFCD00009175
Molecular Formula: C6H13NO2
Molecular Weight: 131.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 40
Flash Point (°C): 32
Specific Gravity (20/20): 0.93
Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: Di-Me-Gly-OEt
Formula:
C₆H₁₃NO₂ MW: 131.17 g/mol Boiling Pt: 40 °C Density: 0.93 (20 °C) Flash Pt: 32 °C |
MDL Number:
MFCD00009175 CAS Number: 33229-89-9 EINECS: 251-411-9 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...